Human recombinant Cystatin F (from cells)
Supplier: ProSci
PRSI91-310EA 761 EUR
PRSI91-310
Human recombinant Cystatin F (from cells)
Proteins and Peptides
CST7 is a secreted protein and primarily expressed in peripheral blood cells and spleen.It is belongs to the cystatin family. The cystatin superfamily encompasses proteins that contain multiple cystatin-like sequences. Some of the members are active cysteine protease inhibitors, while others have lost or perhaps never acquired this inhibitory activity. There are three inhibitory families in the superfamily, including the type 1 cystatins (stefins), type 2 cystatins and the kininogens. The type 2 cystatin proteins are a class of cysteine proteinase inhibitors found in a variety of human fluids and secretions. This gene encodes a glycosylated cysteine protease inhibitor with a putative role in immune regulation through inhibition of a unique target in the hematopoietic system.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag