Human Recombinant NCR3 (from Cells)
Supplier: ProSci Inc.
PRSI91-347EA 505 EUR
PRSI91-347
Human Recombinant NCR3 (from Cells)
Proteins and Peptides
Natural Cytotoxicity Triggering Receptor 3 (NCR3) along with NKp44 and NKp46 constitute a group of receptors termed “Natural Cytotoxicity Receptors”. They play a major role in triggering NK-mediated killing of most tumor cells lines. NKp30 is a type I transmembrane protein having a single extracellular V-like immunoglobulin domain. NKp30 is selectively expressed both in resting and activated human NK cells. In addition, NKp30 is also involved in NK-mediated induction of dendritic cell (DC) maturation. It has been demonstrated that NK cell activation signaling specifically induces lytic activity against several tumor cell types and synthesis of new NF-κB dependent proteins during the initiation of cytotoxicity.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag