Human Recombinant TRAILR3 (from Cells)
Supplier: ProSci Inc.
PRSI91-455EA 270 EUR
PRSI91-455
Human Recombinant TRAILR3 (from Cells)
Proteins and Peptides
Tumor Necrosis Factor Receptor Superfamily Member 10C (TNFRSF10C) is a glycosyl-phosphatidylinositol-linked membrane protein which binds TRAIL with high affinity. TNFRSF10C has the TRAIL-binding extracellular cysteine-rich domains, lacks the intracellular signaling domain. As a result, binding of TRAIL to TRAIL R3 doesn’t transduce an apoptosis signal. The expression of TRAIL R3 gene has been shown to protect cells bearing TRAIL R1 and/or TRAIL R2 from TRAIL-induced apoptosis.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag