Human Recombinant TREM1 (from Cells)
Supplier: ProSci Inc.
PRSI91-457EA 370 EUR
PRSI91-457
Human Recombinant TREM1 (from Cells)
Proteins and Peptides
Triggering Receptor Expressed on Myeloid Cells 1 (TREM-1) is a transmembrane protein with a single Ig-like domain. TREM-1 associates with the adapter protein, DAP12, to deliver an activating signal. TREM-1 is expressed on blood neutrophils and monocytes, and the expression is up-regulated by bacterial LPS. TREM-1 is expressed at high levels on neutrophils of patients with microbial sepsis and in mice with a TREM-1/Fc fusion protein protected mice against LPS-induced shock. Human TREM-1 shares 42% sequence homology with mouse TREM-1.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag