Human Recombinant Poliovirus Receptor (from Cells)
Supplier: ProSci
PRSI91-540EA 455 EUR
PRSI91-540
Human Recombinant Poliovirus Receptor (from Cells)
Proteins and Peptides
Poliovirus Receptor (PVR) is a 70 kDa type I transmembrane single-span glycoprotein that belongs to the nectin-like (Necl) family and was originally identified based on its ability to mediate the cell attachment and entry of poliovirus (PV), an etiologic agent of the central nervous system disease poliomyelitis. PVR contains three Ig-like extracellular domains, a transmembrane segment, and a cytoplasmic tail. The normal cellular function of PVR maybe the involvement of intercellular adhension between epithelial cells. Alternate splicing of the PVR mRNA yields four different isoforms ( alpha, beta , gamma , and delta) with identical extracellular domains.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag