Mouse Recombinant IL11
Supplier: ProSci
PRSI91-599EA 765 EUR
PRSI91-599
Mouse Recombinant IL11
Proteins and Peptides
Interleukin-11(IL-11) is a secreted protein and belongs to the IL-6 superfamily. IL-11 has been demonstrated to improve platelet recovery after chemotherapy-induced thrombocytopenia, induceacute phase proteins, modulate antigen-antibody responses, participate in the regulation of bone cellproliferation and differentiation and could be use as a therapeutic for osteoporosis. IL-11 stimulates the growth of certain lymphocytes and, in the murine model, stimulates an increase in the cortical thickness and strength of long bones. In addition to having lymphopoietic/hematopoietic and osteotrophic properties, it has functions in many other tissues, including the brain, gut, testis and bone.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag