Mouse recombinant Fc gamma RII
Supplier: ProSci
PRSI91-609EA 525 EUR
PRSI91-609
Mouse recombinant Fc gamma RII
Proteins and Peptides
Low affinity immunoglobulin gamma Fc region receptor II (CD32B) is a single-pass type I membrane protein and contains 2 Ig-like C2-type (immunoglobulin-like) domains. The inhibitory CD32B is expressed on B cells and myeloid dendritic cells. Ligation of CD32B on B cells downregulates antibody production and may, in some circumstances, promote apoptosis. Co-ligation of CD32B on dendritic cells inhibits maturation and blocks cell activation. CD32B may also be a target formonoclonal antibody therapy for malignancies.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag