Mouse Recombinant GDF8
Supplier: ProSci
PRSI91-625EA 580 EUR
PRSI91-625
Mouse Recombinant GDF8
Proteins and Peptides
Mouse Growth/differentiation factor 8(Mstn, GDF-8) is a member of the bone morphogenetic protein (BMP) family and the TGF-beta superfamily. This group of proteins is characterized by a polybasic proteolytic processing site which is cleaved to produce a mature protein containing seven conserved cysteine residues. It is expressed specifically in developing and adult skeletal muscle. It exists as a homodimer, and interacts with WFIKKN2, leading to inhibit its activity. This protein can act specifically as a negative regulator of skeletal muscle growth. It regulates cell growth and differentiation in both embryonic and adult tissues. Experiments in mice have improved that GDF-8 is a key regulator of mesenchymal stem cell proliferation and differentiation, and mice lacking Myostatin encoding gene show decreased body fat and a generalized increase in bone density and strength.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag