Human recombinant S100 calcium binding A8 (from E. coli)
Supplier: ProSci
PRSI91-636EA 365 EUR
PRSI91-636
Human recombinant S100 calcium binding A8 (from E. coli)
Proteins and Peptides
Protein S100-A8 (also MRP8 and calgranulin A) is a calcium- and zinc-binding protein.It plays a prominent role in the regulation of inflammatory processes and immune response. S100A8 can induce neutrophil chemotaxis and adhesion. Its role as an oxidant scavenger has a protective role in preventing exaggerated tissue damage by scavenging oxidants. It can act as a potent amplifier of inflammation in autoimmunity as well as in cancer development and tumor spread.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag