Human Recombinant PACSIN2 (from Cells)
Supplier: ProSci
PRSI91-686EA 580 EUR
PRSI91-686
Human Recombinant PACSIN2 (from Cells)
Proteins and Peptides
Protein Kinase C and Casein Kinase Substrate in Neurons Protein 2 (PACSIN2) is a member of the PACSIN family. PACSIN2 is localized to the plasma membrane via its coiled-coil domain. PACSIN2 is widely expressed and contains one FCH domain and one SH3 domain. PACSIN2 forms homo- and hetero-aggregates with other PACSINs. PACSIN2 may play a role in vesicle formation and transport. In addition, PACSIN2 is involved in linking the actin cytoskeleton with vesicle formation by regulating tubulin polymerization.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag