Human recombinant Calmegin (from Cells)
Supplier: ProSci
PRSI91-690EA 761 EUR
PRSI91-690
Human recombinant Calmegin (from Cells)
Proteins and Peptides
Calmegin (CLGN) is a member of the calreticulin family. Calmegin is a testis-specific endoplasmic reticulum chaperone protein. Calmegin binds calcium ions and interacts with PDILT. Calmegin may play a role in spermatogeneisis and infertility.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag