Human Recombinant HDGFRP3 (from Cells)
Supplier: ProSci
PRSI91-693EA 580 EUR
PRSI91-693
Human Recombinant HDGFRP3 (from Cells)
Proteins and Peptides
Hepatoma-Derived Growth Factor-Related Protein 3 (HDGFRP3) belongs to the HDGF family. HDGFRP3 can be found in testis, heart, spinal cord and brain. HDGFRP3 localizes to the nucleus and contains one PWWP domain. HDGFRP3 enhances DNA synthesis and may have a role in cell proliferation.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag