Human Recombinant FLRT3 (from Cells)
Supplier: ProSci
PRSI91-706EA 455 EUR
PRSI91-706
Human Recombinant FLRT3 (from Cells)
Proteins and Peptides
Leucine-Rich Repeat Transmembrane Protein FLRT3 (FLRT3) is a member of the fibronectin leucine rich transmembrane protein (FLRT) family. Proteins in this family play an role in cell adhesion and/or receptor signalling. FLRT3 is a single-pass type I membrane protein and contains one fibronectin type-III domain, ten LRR (leucine-rich) repeats, one LRRCT domain, and one LRRNT domain. FLRT3 may have a function in cell adhesion and/or receptor signaling. FLRT3 may regulate cellular adhesion between the epithelial apical ridge and the underlying mesenchyme and in establishing the dorso-ventral position of the ridge.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag