Human Recombinant SH3KBP1 (from Cells)
Supplier: ProSci
PRSI91-709EA 580 EUR
PRSI91-709
Human Recombinant SH3KBP1 (from Cells)
Proteins and Peptides
SH3 Domain-Containing Kinase-Binding Protein 1 (SH3KBP1) contains 3 SH3 domains. SH3KBP1 is widely expressed in many tissues, it is also expressed in some cancer cell lines. SH3KBP1 plays a role in the regulation of endocytosis and lysosomal degradation of ligand-induced receptor tyrosine kinases. SH3KBP1 is involved in regulation of ligand-dependent endocytosis of the IgE receptor and in the regulation of cellular stress response via the MAPK pathways. SH3KBP1 is also required for the control of cell shape and migration.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag