Human Recombinant Neurotrimin (from Cells)
Supplier: ProSci
PRSI91-793EA 580 EUR
PRSI91-793
Human Recombinant Neurotrimin (from Cells)
Proteins and Peptides
Neurotrimin localizes to the cell membrane and contains three Ig-like C2-type (immunoglobulin-like) domains. Neurotrimin acts as a glycosylphosphatidylinositol (GPI)-anchored neural cell adhesion molecule. Neurotrimin may promote neurite outgrowth and adhesion via homophilic and heterophilic interactions.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag