Human Recombinant Nucleotide exchange factor SIL1 (from Cells)
Supplier: ProSci
PRSI91-845EA 455 EUR
PRSI91-845
Human Recombinant Nucleotide exchange factor SIL1 (from Cells)
Proteins and Peptides
Nucleotide exchange factor SIL1, also named BiP-associated protein, is a member of the SIL1 family. It is a resident endoplasmic reticulum (ER), N-linked glycoprotein with an N-terminal ER targeting sequence, 2 putative N-glycosylation sites, and a C-terminal ER retention signal. It is highly expressed in tissues which produce large amounts of secreted proteins such as kidney, liver and placenta. This protein functions as a nucleotide exchange factor for another unfolded protein response protein. Mutations in this gene have been associated with Marinesco-Sjogren syndrome.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag