Human recombinant CDNF (from cells)
Supplier: ProSci
PRSI91-877EA 525 EUR
PRSI91-877
Human recombinant CDNF (from cells)
Proteins and Peptides
erebral Dopamine Neurotrophic Factor (CDNF), also known as ARMETL1 (ARMET-like protein 1), is a secreted protein with eight conserved cysteine residues.It is belongs to the ARMET family. CDNF/ARMETL1 is a evolutionary conserved protein which can protect and restore the function of dopaminergic neurons in the rat model of Parkinson's disease, suggesting that CDNF might be beneficial for the treatment of Parkinson's disease. CDNF is widely expressed in neurons in several brain regions including cerebral cortex, hippocampus, substantia nigra, striatum and cerebellum. Human CDNF is glycosylated and secreted from transiently transfected cells. CDNF promotes the survival, growth, and function of dopamine-specific neurons and is expressed in brain regions that undergo cocaine-induced neuroplasticity.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag