Human recombinant CXCL10 (from cells)
Supplier: ProSci
PRSI91-905EA 553 EUR
PRSI91-905
Human recombinant CXCL10 (from cells)
Proteins and Peptides
Human C-X-C Motif Chemokine Ligand 10 (CXCL10) is a non-ELR chemokine secreted by various cell types, such as monocytes, endothelial cells and fibroblasts, in response to IFN- gamma . CXCL10 functions via chemokine receptor CXCR3. CXCL10 has been attributed to several roles, such as chemoattraction for activated T-lymphocytes, inhibition of angiogenesis, and antitumor activity.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-Fc-6 His tag