Human recombinant SCF (from cells)
Supplier: ProSci
PRSI91-924EA 553 EUR
PRSI91-924
Human recombinant SCF (from cells)
Proteins and Peptides
Stem Cell Factor (SCF) is a hematopoietic growth factor that exerts its activity at the early stages of hematopoiesis. SCF stimulates the proliferation of myeloid, erythroid, and lymphoid progenitors in bone marrow cultures and has been shown to act synergistically with colony stimulating factors.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag