Mouse Recombinant PSGL-1
Supplier: ProSci
PRSI91-961EA 395 EUR
PRSI91-961
Mouse Recombinant PSGL-1
Proteins and Peptides
P-selectin glycoprotein ligand 1 is the high affinitycounter-receptor for P-selectin on expressed on activated endothelial cells and platelets. As such, it plays a critical role in the tethering of these cells to activated platelets or endothelia expressing P-selectin. As cell adhesion molecules, multiple studies have shown that PSGL-1/ P-selectin interaction is required for the normal recruitment of leukocytes during inflammatory reactions, and also participates in hemostatic responses. PSGL-1 can also bind to other two members of the selectin family, E-selectin (endothelial) and L-selectin (leukocyte), but binds best to P-selectin.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-Fc tag