Human Recombinant BDH2 (from E. coli)
Supplier: ProSci
PRSI91-973EA 610 EUR
PRSI91-973
Human Recombinant BDH2 (from E. coli)
Proteins and Peptides
3-Hydroxybutyrate Dehydrogenase Type 2 belongs to the short-chain dehydrogenases/reductases (SDR) family. 3-Hydroxybutyrate Dehydrogenase Type 2 may play an important role in the peripheral utilization of 3-hydroxybutyrate. The cytoplasmic localization with its high ratio of oxidized NAD+, the NAD+ dependence and the kinetic parameters of 3-Hydroxybutyrate Dehydrogenase Type 2 make it suitable to conbert high levels of circulating 3-hydroxybutyrate into acetoacetate.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag