Human Recombinant PI-TP-alpha (from E. coli)
Supplier: ProSci
PRSI91-977EA 610 EUR
PRSI91-977
Human Recombinant PI-TP-alpha (from E. coli)
Proteins and Peptides
Phosphatidylinositol Transfer Protein alpha Isoform (PITPNA) is found in the cytoplasm and belongs to the PtdIns transfer protein family. PITPNA is a ubiquitous and highly conserved protein in multicellular eukaryotes that catalyzes the exchange of phospholipids between membranes and participates in cellular phospholipid metabolism, signal transduction and vesicular trafficking in vivo. It is expressed in a wide range of tissues and implicated in phospholipase C signaling and in the production of phosphatidylinositol 3, 4, 5-trisphosphate (PIP3) by phosphoinositide-3-kinase.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag