Human Recombinant CDC73 (from E. coli)
Supplier: ProSci
PRSI92-011EA 455 EUR
PRSI92-011
Human Recombinant CDC73 (from E. coli)
Proteins and Peptides
Parafibromin is expressed in the adrenal and parathyroid glands, kidney, and heart. As a tumor supressor, Parafibromin may be involved in transcriptional and post-transcriptional control pathways, also through the regulation of cyclin D1/PRAD1 expression, involved the cell cycle progression. Parafibromin is a component of the the PAF protein complex and interacts with a Set1-like complex that has histone methyltransferase activity and methylates histone H3. Defects in Parafibromin can cause hyperparathyroidism-jaw tumor syndrome, familial isolated hyperparathyroidism, and parathyroid carcinoma.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag