Human recombinant Carboxypeptidase A4 (from HEK293 cells)
Supplier: ProSci
PRSI96-198EA 480 EUR
PRSI96-198
Human recombinant Carboxypeptidase A4 (from HEK293 cells)
Enzymes
Carboxypeptidases are zinc-containing exopeptidases that catalyze the release of carboxy-terminal amino acids, and are synthesised as zymogens that are activated by proteolytic cleavage. Carboxypeptidases cleave amino acids from the C-terminus of proteins and peptides and many are metalloproteases. Carboxypeptidase A4 (CPA4) is also known as Carboxypeptidase A3 (CPA3), which belongs to the peptidase M14 family. Fetal express the protein in the adrenal gland, brain, heart, intestine, kidney, liver and lung.