Human recombinant Dipeptidyl Aminopeptidase Ii Prot (from HEK293 cells)
Supplier: ProSci
PRSI96-247EA 455 EUR
PRSI96-247
Human recombinant Dipeptidyl Aminopeptidase Ii Prot (from HEK293 cells)
Dipeptidyl peptidase 2 (DPP2 or DPP II) is also known as Dipeptidyl peptidase 7 (DPP7), Quiescent cell proline dipeptidase (QPP), which belongs to the peptidase S28 family. DPP7 / DPP2 is detected in seminal plasma (at protein level). When the pH is 5.5, Dipeptidyl peptidase 2 display a optimum catalytic activity. DPP7 has low activity at pH 7 and is inactive at pH 8. DPP7 plays an important role in the degradation of some oligopeptides.