Human Recombinant Leukotriene A4 Hydrolase (from HEK293 cells)
Supplier: ProSci
PRSI96-524EA 455 EUR
PRSI96-524
Human Recombinant Leukotriene A4 Hydrolase (from HEK293 cells)
Enzymes
Leukotriene A4 hydrolase (LTA4H) is a protein which belongs to the peptidase M1 family, specifically those acting on ether bonds (ether hydrolases). LTA4H is bifunctional enzyme which converts leukotriene A4 to leukotriene B4 and acts as an aminopeptidase. LTA4H participates in arachidonic acid metabolism. LTA4H is inhibited by bestatin and subject to suicide inhibition by leukotriene A4, due to the formation of a covalent adduct at Tyr-379. LTA4H binds 1 zinc ion per subunit.