Human recombinant ribonuclease Pancreatic (from cells)
Supplier: ProSci
PRSI91-739EA 658 EUR
PRSI91-739
Human recombinant ribonuclease Pancreatic (from cells)
Enzymes
Ribonuclease Pancreatic is a secreted enzyme that belongs to the pancreatic ribonuclease family. RNASE1 is an endonuclease that cleaves internal phosphodiester RNA bonds on the 3'-side of pyrimidine bases. RNASE1 prefers poly(C) as a substrate and hydrolyses 2',3'-cyclic nucleotides, with a pH optimum near 8.0. RNASE1 acts on single stranded and double stranded RNA.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.