Human recombinant Inosine Triphosphatase (from E. coli)
Supplier: ProSci
PRSI91-989EA 789 EUR
PRSI91-989
Human recombinant Inosine Triphosphatase (from E. coli)
Enzymes
Inosine Triphosphate Pyrophosphatase (ITPase) is a cytoplasmic enzyme that belongs to the HAM1 NTPase family. ITPase hydrolyses the non-canonical purine nucleotides inosine triphosphate (ITP) and deoxyinosine triphosphate (dITP) to the monophosphate nucleotide (IMP) and diphosphate. The ITPase enzyme acts as a homodimer and does not distinguish between the deoxy- and ribose forms. ITPase probably excludes non-canonical purines from RNA and DNA precursor pools, thus preventing their incorporation into RNA and DNA and avoiding chromosomal lesions. Defects in ITPase is thought to be inherited and is characterised by an over-accumulation of ITP in erythocytes, leukocytes and fibroblasts.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.