Human Recombinant Vaccinia-related kinase 3 (from Cells)
Supplier: ProSci
PRSI92-347EA 610 EUR
PRSI92-347
Human Recombinant Vaccinia-related kinase 3 (from Cells)
Enzymes
Inactive serine/threonine-protein kinase VRK3 is a 474 amino acids protein that belongs to the protein kinase superfamily, CK1 Ser/Thr protein kinase family and VRK subfamily. It contains a protein kinase domain. VRK3 is widely expressed in human tissues and the protein localises to the nucleus. VRK3 regulates several transcription factors, nuclear envelope assembly, and chromatin condensation and is also required for cell cycle progression .
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.