Whole Blood, prostate cancer (PC)
HCMSPB000F-2-PCEA 0 EUR
HCMSPB000F-2-PC
Whole Blood, prostate cancer (PC)
Whole blood is used for researching the causes, diagnosis, supportive care and treatment of prostate cancer (PC). Collected from IRB-consented donors diagnosed with with prostate cancer.
No pre-screening is done on fresh products, but donors are tested on day of collection for Hepatitis B, Hepatitis C, and HIV 1/2, HTLV-I/II, HIV-1/HCV/HBV nucleic acid, WNV nucleic acid, Trypanasoma cruzi and Syphilis.
Storage: Use immediately upon receipt.
Caution: Intended for research use only and not for direct therapeutic or diagnostic use in human or animals, or for further manufacturing of therapeutic products or devices.