Human serum, prostate cancer
HCMSPB007C-1-PCEA 0 EUR
HCMSPB007C-1-PC
Human serum, prostate cancer
Human serum used for researching the causes, diagnosis, supportive care and treatment of prostate cancer (PC). Collected from IRB-consented donors diagnosed with PC.
This prostate cancer is the most commonly found in men, treatment for this cancer is in active research development stage, where improvements are being made among many standard prostate cancer treatment methods.
No prescreening on fresh products, but donors will be tested on day of collection for Hepatitis B, Hepatitis C, HIV 1/2, HTLV-I/II, HIV-1/HCV/HBV nucleic acid, WNV nucleic acid, Trypanasoma cruzi and Syphilis.
Storage: Store at −80 °C.
Caution: Intended for research use only and not for direct therapeutic or diagnostic use in human or animals, or for further manufacturing of therapeutic products or devices.