Sodium orthovanadate ≥99%, solid

Supplier: BioVision

BIOV9470-1GEA 65.4 EUR
BIOV9470-1G BIOV9470-5G
Sodium orthovanadate ≥99%, solid
Sodium orthovanadate

Sodium orthovanadate is a commonly used general inhibitor for protein tyrosine phosphatases (PTPs). It is a competitive inhibitor. The inhibition by Vanadate is completely reversible upon the addition of EDTA or by dilution.


Caution: Protect from air and moisture

Formula: Na₃O₄V
MW: 183,91 g/mol
MDL Number: MFCD00003511
CAS Number: 13721-39-6

Order Now

Specification Test Results

Appearance White to off-white solid
Solubility H₂O (~50 mg/ml)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR