BIOV2280
BIOV2280-5EA 198 EUR
BIOV2280-5 BIOV2280-25
BIOV2280
Delamanid is a antituberculosis drug that inhibits mycolic acid synthesis and has shown potent in vitro and in vivo activity against drug-resistant strains of Mycobacterium tuberculosis.
  • White solid
  • Soluble in DMSO
Caution: For research use only. Not for use in humans.