Diethyl phosphate 96%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
diethyl hydrogen-phosphoric acid
, di-Ethyl phosphate
, Diethylphosphoric acid
, Diethyl hydrogen phosphate
ACRO407420050EA
129
EUR
ACRO407420050
Diethyl phosphate 96%
Diethyl phosphate
Formula:
C₄H₁₁O₄P MW: 154,102 g/mol Boiling Pt: 203 °C (760 mmHg) Melting Pt: 6 °C Density: 1,29 g/cm³ Flash Pt: 91 °C Storage Temperature: Ambient |
MDL Number:
MFCD00039889 CAS Number: 598-02-7 UN: 3265 ADR: 8,III |
Specification Test Results
Appearance | Clear colorless light yellow Liquid |
Infrared spectrum | Conforms |
Titration with NaOH | ≥95 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...