1,2-Diiodotetrafluorobenzene 99%

Warning

ACRO311690050EA 227 EUR
ACRO311690050 ACRO311690010
1,2-Diiodotetrafluorobenzene 99%
1,2-Diiodotetrafluorobenzene
Formula: C₆F₄I₂
MW: 401,87 g/mol
Melting Pt: 49…50 °C
Storage Temperature: Ambient
MDL Number: MFCD00019011
CAS Number: 2708-97-6

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 49 to 53 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR