SeQuant® CARS™ and SAMS™, IC suppressor system
1.50616.0100EA 213 EUR
MERC1.50616.0100 MERC1.50611.0001
SeQuant® CARS™ and SAMS™, IC suppressor system
Determination of anions by ion chromatography is now easy. Simply use the continuous background conductivity suppression system SeQuant® CARS™, equipped with the membrane suppressor SeQuant® SAMS™, to enhance the results.
  • Low and stable background levels and high sensitivity in routine analysis
  • Can be integrated with any ion chromatography system
  • Suitable also for high ion-strength eluents and gradient applications
SeQuant® CARS™ is a continuous chemical regeneration system for membrane suppressor like the SeQuant® SAMS™. SeQuant® SAMS™ is a chemically-regenerated membrane suppressor for anion chromatography. Its operation is based on selective exchange of protons (H+) from an external regeneration channel, for cations (e.g., Na+) from the eluent.