SV40, BioProbe®
Supplier: Enzo Life Sciences
ALEXENZ-40845EA 388 EUR
ALEXENZ-40845
SV40, BioProbe®
Vectors, Plasmids and Libraries
The SV40 BioProbe® labeled probe is prepared by nick translation of the entire genome of SV40, a member of the papovavirus family. SV40 has served as a model virus for studies of transformation and gene regulation. Fragment size range: 100 to 1000 base pairs (as estimated by agarose gel electrophoresis).
Concentration: 20 µg/ml.
Size: 2 µg.