Hygromycin B (from Streptomyces hygroscopicus) ≥80% (by HPLC), liquid
Supplier: Enzo Life Sciences
ALEXALX380059UM01EA
239
EUR
ALEXALX380059UM01
Hygromycin B (from Streptomyces hygroscopicus) ≥80% (by HPLC), liquid
Hygromycin B
Antibiotic and antiviral.
Antibiotic with high activity. Inhibits growth of prokaryotic and eukaryotic microorganisms and mammalian cells. Inhibits protein synthesis at the translocation step on the 70S ribosome and causes misreading of the mRNA. Penetrates cells that have been permeabilized by virus infection, hence it can act as an effective antiviral agent.
Dark brown solution
Packaging: Shipping temperature: Ambient
Formula:
C₂₀H₃₇N₃O₁₃ MW: 527,5 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD06795479 CAS Number: 31282-04-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...