Brain (mouse) tissue extract
Supplier: Enzo Life Sciences
ALEXADILYTMB100FEA 165 EUR
ALEXADILYTMB100F
Brain (mouse) tissue extract
Specimens Preserved Specimens Preserved Animal Materials
  • Liquid
Packaging: In SDS-PAGE sample buffer (pH: 6,8) containing 40 mM TRIS-HCl, 1% SDS, 50 mM DTT, 7,5% glycerol and 0,003% bromophenol blue.