Liver (rat) microsome extract
Supplier: Enzo Life Sciences
ALEXADILYTRM100FEA 161 EUR
ALEXADILYTRM100F
Liver (rat) microsome extract
Specimens Preserved Specimens Preserved Animal Materials
Liquid extract in a SDS-PAGE sample buffer (pH: 6,8) containing 40 mM TRIS-HCl, 1% SDS, 50 mM DTT, 7,5% glycerol and 0,003% bromophenol blue.