4-(Dimethylamino)-N-[7-(hydroxyamino)-7-oxoheptyl]benzamide ≥98% (by HPLC)
Supplier: Enzo Life Sciences
Synonyms:
4-(Dimethylamino)-N-(7-(hydroxyamino)-7-oxoheptyl)benzamide
, M 344
ALEXALX270297M001EA
207
EUR
ALEXALX270297M001
ALEXALX270297M005
4-(Dimethylamino)-N-[7-(hydroxyamino)-7-oxoheptyl]benzamide ≥98% (by HPLC)
4-(Dimethylamino)-N-[7-(hydroxyamino)-7-oxoheptyl]benzamide
Potent inhibitor of histone deacetylases (HDACs) (IC50≤1µM).
- HDAC inhibitor
White crystalline solid.
Soluble in DMSO (1mg/ml)
Formula:
C₁₆H₂₅N₃O₃ MW: 307,39 g/mol |
MDL Number:
MFCD03453554 CAS Number: 251456-60-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...