1,1-Dimethylbiguanide hydrochloride ≥97%
Supplier: Enzo Life Sciences
Synonyms:
AMPK Signaling Activator VII
, Metformin
, 1,1-Dimethylbiguanide Hydrochloride
, Metformin hydrochloride
ALEXALX270432G005EA
62.1
EUR
ALEXALX270432G005
1,1-Dimethylbiguanide hydrochloride ≥97%
1,1-Dimethylbiguanide hydrochloride
Antidiabetic agent
Metformin is an antidiabetic agent. Reduces the cardiovascular complications of diabetes. Does not induce hypoglycemia. Inhibits hepatic gluconeogenesis via activation of the LKB1/AMPK pathway. Has antiproliferative effects in cancer cell lines.
Formula:
C₄H₁₂ClN₅ MW: 165,625 g/mol |
MDL Number:
MFCD00012582 CAS Number: 1115-70-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...