Strobilurin B

ALEXALX380144M001EA 567 EUR
ALEXALX380144M001
Strobilurin B
Strobilurin B

Antifungal agent


Antifungal agent and respiration inhibitor. Acts by inhibiting mitochondrial respiration in fungi. Binds at the Q0-centre on cytochrome b and blocks the electron transfer between cytochrome b and cytochrome c1.


Isolated from Strobilurus sp.

Formula: C₁₇H₁₉ClO₄
MW: 322,8 g/mol
CAS Number: 65105-52-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR