Forskolin ≥99% (by TLC)

Supplier: Enzo Life Sciences

ALEXBML-CN100-0010EA 162 EUR
ALEXBML-CN100-0010 ALEXBML-CN100-0100
Forskolin ≥99% (by TLC)
Forskolin

Adenylate cyclase activator


Activates adenylate cyclase by directly interacting with the catalytic unit of the enzyme leading to an increase in the intracellular concentration of cAMP. Widely used tool for the investigation of the role of cAMP as a second messenger with a broad range of potential therapeutic applications. Inotropic agent and vasodilator. Induces platelet aggregation. Inhibits ion channels by a mechanism that does not involve cAMP. Non-competitive inhibitor of nicotinic acetylcholine receptors.


Isolated from Coleus forskohlii.

Formula: C₂₂H₃₄O₇
MDL Number: MFCD00082317
CAS Number: 66575-29-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR