3'-Carbamoyl-[1,1'-biphenyl]-3-ylcyclohexylcarbamate ≥98% (by TLC)

Supplier: Enzo Life Sciences

Synonyms: URB597

ALEXBML-EI327-0025EA 249 EUR
ALEXBML-EI327-0025
3'-Carbamoyl-[1,1'-biphenyl]-3-ylcyclohexylcarbamate ≥98% (by TLC)
3'-Carbamoyl-[1,1'-biphenyl]-3-ylcyclohexylcarbamate

URB-597 is a potent and selective inhibitor of the enzyme fatty acid amide hydrolase (FAAH), which is involved in the hydrolysis of endogenous cannabinoids such as anandamide. Inhibition of FAAH may serve to enhance the analgesic effects of endogenous cannabinoids. URB-597 has been shown to inhibit FAAH in rat cortical neuron membranes and in intact neurons with IC50 values of 4.6nM and 0.5nM, respectively. The compound has also been shown to have anxiolytic effects in rats (ID50=0.15mg/kg). Because URB-597 acts on FAAH and does not interfere with the binding of anandamide to the CB1 and CB2 receptors, it is a novel tool for drug development.


  • FAAH inhibitor


White solid.


Soluble in 100% ethanol (1mg/ml), dimethyl formamide (10mg/ml) or DMSO (10mg/ml).

Formula: C20H22N2O3
MW: 338,4 g/mol
Melting Pt: 155…157 °C
CAS Number: 546141-08-6
https://ie.vwr.com/stibo/web/std.lang.all/37/75/45403775.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR