5,8,11,14-Eicosatetraynoic acid ≥98%

Supplier: Enzo Life Sciences

Synonyms: ETYAOctadehydroarachidonic acid

ALEXBML-ET004-0020EA 107 EUR
ALEXBML-ET004-0020 ALEXBML-ET004-0100
5,8,11,14-Eicosatetraynoic acid ≥98%
5,8,11,14-Eicosatetraynoic acid

Arachidonic acid uptake inhibitor


Inhibits arachidonic acid uptake. Inhibits cyclooxygenase (ID50=8 µM) and all lipoxygenases (ID50=10, 0.3, and 0.2 µM for 5-, 12- and 15-LO respectively) in whole cells. Inhibits PLA2 and cytochrome P-450. Modulates Ca2+ entry into cells. Stimulates luteinizing hormone release from cultured pituitary cells. PPAR agonist.

Formula: C₂₀H₂₄O₂
MDL Number: MFCD00036967
CAS Number: 1191-85-1
https://ie.vwr.com/stibo/web/std.lang.all/38/32/45403832.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR