1-(3-Chloroanilino)-4-phenylphthalazine ≥98% (by TLC)

Supplier: Enzo Life Sciences

Synonyms: MY-54451-(3-Chlorophenyl)amino-4-phenylphthalazine

ALEXBML-PD155-0010EA 260 EUR
ALEXBML-PD155-0010
1-(3-Chloroanilino)-4-phenylphthalazine ≥98% (by TLC)
1-(3-Chloroanilino)-4-phenylphthalazine

PDE inhibitor


Selective inhibitor of cGMP-specific phosphodiesterase (PDE V, IC50=0.6µM).

Formula: C₂₀H₁₄ClN₃
MW: 331,80 g/mol
MDL Number: MFCD00209838
CAS Number: 78351-75-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR