Prostaglandin E1 ≥99%
Supplier: Enzo Life Sciences
Synonyms:
PGE1
, 7-((1R,2R,3R)-3-Hydroxy-2-((S,E)-3-hydroxyoct-1-en-1-yl)-5-oxocyclopentyl)heptanoic acid
, Alprostadil
, Caverject
ALEXBML-PG006-0010EA
235
EUR
ALEXBML-PG006-0010
Prostaglandin E1 ≥99%
Prostaglandin E1
Prostanoid receptor ligand
Biosynthesis from dihomo-gamma-linolenic acid. A major prostaglandin in semen. Inhibits human platelet aggregation. Effects induced by receptor-mediated elevation of cAMP.
Formula:
C₂₀H₃₄O₅ MW: 354,487 g/mol |
MDL Number:
MFCD00077860 CAS Number: 745-65-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...