2-Methoxyestradiol

Supplier: Enzo Life Sciences

ALEXBML-S540-0050EA 388 EUR
ALEXBML-S540-0050 ALEXBML-S540-0010
2-Methoxyestradiol
2-Methoxyestradiol

Estrogen receptor inhibitor


An endogenous estrogen metabolite with low affinity for the estrogen receptor but which disrupts microtubule function. Potent inhibitor of endothelial cell proliferation and migration. Inhibits bFGF and VEGF-induced corneal neovascularization in mice. Posttranscriptionally downregulates HIF-1alpha expression.

Formula: C₁₉H₂₆O₃
MDL Number: MFCD00010489
CAS Number: 362-07-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR