Zirconium(IV) oxide 20% in water, colloidal dispersion
|
Danger
|
Synonyms:
Zirconium dioxide
, Zirconium Oxide
40124.A4EA
280
EUR
40124.A4
Zirconium(IV) oxide 20% in water, colloidal dispersion
Zirconium (IV) oxide
Formula:
ZrO₂ MW: 123,22 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011310 CAS Number: 1314-23-4 EINECS: 215-227-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...